Learn More
Antimony(III) Fluoride, Sigma-Aldrich™
Product Code. 15869530
Change view
Click to view available options
Quantity:
5 g
250 g
Unit Size:
250g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15869530 250 g 250g
15859530 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15869530 Supplier Sigma Aldrich Supplier No. 8078390250
 Restricted Item
Estimated Shipment: 26-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7783-56-4
Grade Research
Molecular Weight (g/mol) 178.76
Specifications
Boiling Point 376°C/1013 hPa
Melting Point 292°C
Quantity 250 g
Density 4.38 g/cm3 at 20°C
MDL Number MFCD00011218
Synonym Antimony(III) fluoride, Antimony trifluoride, Antimony(III) fluoride
Chemical Name or Material Antimony(III) Fluoride