missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III) chloride, Sigma-Aldrich™

Product Code. 32238080
Change view
Click to view available options
Quantity:
45 kg
Unit Size:
45kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32238080 45 kg 45kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32238080 Supplier Sigma Aldrich Supplier No. 8146569045
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

For synthesis

TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 233°C (1013 hPa)
Chemical Name or Material Antimony(III) chloride
Melting Point 73°C
CAS 10025-91-9
Quantity 45 kg
Density 3.14 g/cm³ at 20°C
Synonym Antimony(III) chloride, Antimonous acid, Antimony trichloride, Antimony(III) chloride
Molecular Weight (g/mol) 228.12
Grade Not Available
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.