missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Antiblaze V6-d16, TRC
Specifications
Specifications
| Chemical Name or Material | Antiblaze V6-d16 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C13 D16 H8 Cl6 O8 P2 |
| Quantity | 1 mg |
| Synonym | P,P'-[2,2-bis(chloromethyl)-1,3-propanediyl] Phosphoric Acid P,P,P',P'-tetrakis(2-chloroethyl) Ester-d16, 2,2-bis(Chloromethyl)-1,3-propanediyl tetrakis(2-chloroethyl) Ester Phosphoric Acid-d16, bis(2-Chloroethyl) Ester Phosphoric Acid Diester with 2,2-bis(Chloromethyl)-1,3-propanediol-d16, 2,2-Bis(chloromethyl)-1,3-propanediol 2-Chloroethanol Phosphate (1:4:2)-d16, 2,2-Bis(chloromethyl)-1,3-propanediol Bis[bis(2-chloroethyl) Phosphate]-d16, 2,2-Bis(chloromethyl)-1,3-propanediyl Bis[bis(2-chloroethyl) Phosphate]-d16, Amgard V 6-d16, Antiblaze 100-d16, Antiblaze AB 100-d16, Antiblaze V 6-d16, Phosgard 2XC20-d16 |
| InChI Formula | InChI=1S/C13H24Cl6O8P2/c14-1-5-22-28(20,23-6-2-15)26-11-13(9-18,10-19)12-27-29(21,24-7-3-16)25-8-4-17/h1-12H2/i1D2,2D2,3D2,4D2,5D2,6D2,7D2,8D2 |
| SMILES | [2H]C([2H])(Cl)C([2H])([2H])OP(=O)(OCC(CCl)(CCl)COP(=O)(OC([2H])([2H])C([2H])([2H])Cl)OC([2H])([2H])C([2H])([2H])Cl)OC([2H])([2H])C([2H])([2H])Cl |
| IUPAC Name | [2-[bis(2-chloro-1,1,2,2-tetradeuterioethoxy)phosphoryloxymethyl]-3-chloro-2-(chloromethyl)propyl] bis(2-chloro-1,1,2,2-tetradeuterioethyl) phosphate |
| Molecular Weight (g/mol) | 599.089 |
| Formula Weight | 596.008 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?