Learn More
Clark Scientific™ Polystyrene Anti-static Spill Trays
Product Code. 12928837
Change view
Click to view available options
Dimensions (L x W):
475 x 360 mm
Unit Size:
Pack of 25
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
12928837 475 x 360 mm Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12928837 Supplier Clark Scientific™ Supplier No. AT1PK25
  • ONLINE EXCLUSIVE PRICE
    81.70 EUR
    74.23 EUR / Pack of 25
    Save 7.47 EUR
    9% Off
View all promotions
Estimated Shipment: 09-12-2026
Made from anti-static, high impact polystyrene, these spill trays are ideal in most research, scientific, laboratory and other environments.
  • Made from anti-static, high impact polystyrene
  • Sufficient depth to safely contain even the largest liquid spills
  • Wipe clean for repeated use
Specifications
Dimensions (L x W) 475 x 360 mm