Learn More
BD anti-Rat Ig, k Light Chain Magnetic Particles - DM IMag-DM, Clone: MRK-1
Product Code. 15839158
Click to view available options
Quantity:
5 mL
Unit Size:
Each
Product Code. 15839158
Brand: BD 558002
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Cell Separation
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store undiluted at 4°C. Protect from prolonged exposure to light. Do not freeze.
Quantity 5 mL
For Use With (Application) Cell separation