Learn More
PPP2R2B, Human, IP-WB Antibody Pair (PW1), Abnova™
Product Code. 16098148
Change view
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16098148 1 Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16098148 Supplier Abnova Supplier No. H00005521PW1.1Set
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
Includes
Antibody pair for IP: mouse monoclonal anti-PPP2R2B (300μg), Antibody pair for WB: rabbit polyclonal anti-PPP2R2B (50μL)
For immunoprecipitation and to detect the precipitated protein in western blot.
Specifications
Antigen PPP2R2B
Applications Immunoprecipitation, Western Blot
Classification Monoclonal/Polyclonal
Conjugate Unconjugated
Description This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
Gene PPP2R2B
Gene Alias B55-BETA/FLJ95686/MGC24888/PP2A-B55BETA/PP2A-PR55B/PP2AB-BETA/PP2APR55-BETA/PR2AB-BETA/PR2AB55-BETA/PR2APR55-BETA/PR52B/PR55-BETA/SCA12
Gene Symbols PPP2R2B
Host Species Mouse, Rabbit
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5521
Includes Antibody pair set content: 1. Antibody pair for IP: mouse monoclonal anti-PPP2R2B (300 ug) 2. Antibody pair for WB: rabbit polyclonal anti-PPP2R2B (50 ul)
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Form Liquid
Show More Show Less
For Research Use Only