Learn More
Ambion™ Anti-miR™ miRNA Inhibitor
Product Code. 10680026
Change view
Click to view available options
Quantity:
20 nmol
5 nmol
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10680026 20 nmol Each
10299484 5 nmol Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10680026 Supplier Ambion™ Supplier No. AM17001
Anti-miR™ miRNA Inhibitor
miRNA inhibitors are small, chemically modified single-stranded RNA molecules designed to specifically bind to and inhibit endogenous miRNA molecules and enable miRNA functional analysis by down-regulation of miRNA activity.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Room temperature
Description Custom Anti-miR™ miRNA Inhibitor, 20nmol, Powder, Small, Chemically Modified Single-Stranded RNA Molecules, Designed to Specifically Bind to and Inhibit Endogenous miRNA, Molecules and Enable miRNA Functional Analysis by Down-Regulation of miRNA Activity
Product Type Custom Anti-miR™ miRNA Inhibitor
Quantity 20 nmol