Learn More
BD anti-Ly-6G and Ly-6C (Gr-1) Magnetic Particles - DM IMag-DM, Clone: RB6-8C5
Product Code. 15819218
missing translation for 'orderingAttributeHoverText'
Quantity:
10 mL
missing translation for 'unitSize'
Each
Product Code. 15819218
missing translation for 'mfr': BD 558111
Cell Separation
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store undiluted at 4°C.
Quantity 10 mL
For Use With (Application) Cell Separation