Learn More
BD anti-CD45RO Magnetic Particles - DM IMag-DM, Clone: UCHL1
Click to view available options
Quantity:
5 mL
Unit Size:
Each
Description
Cell Separation
Specifications
Specifications
| Content And Storage | Store undiluted at 4°C. |
| Quantity | 5 mL |
| For Use With (Application) | Cell separation |