missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Beta-casein Monoclonal Antibody (F20.14)

Description
Suggested positive control: human lactating mammary glands.
This gene is a member of the beta casein family. There are two types of casein protein, beta (encoded by this gene) and kappa, both of which are secreted in human milk. Beta casein is the principal protein in human milk and the primary source of essential amino acids for a suckling infant. Beta and kappa casein proteins acting together form spherical micelles which bind within them important dietary minerals, such as calcium and phosphorous. Alternative splicing results in multiple transcript variants.
Specifications
Specifications
| Antigen | Beta-casein |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | F20.14 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | CSN2 |
| Gene Accession No. | P05814 |
| Gene Alias | beta casein; beta-caesin; beta-casein; beta-casein A3; beta-casein precursor; CASB; casein beta; CSN2; kappa-casein; Unknown (protein for MGC:133765); unnamed protein product |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?