missing translation for 'onlineSavingsMsg'
Learn More

Anthrone, MP Biomedicals™

Product Code. 11487780 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11487780 100 g 100g
11467780 10 g 10g
11477780 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11487780 Supplier MP Biomedicals Supplier No. 0210069180
  • ONLINE EXCLUSIVE PRICE
    427.00 EUR
    406.35 EUR / 100g
    Save 20.65 EUR
    4% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 90-44-8
Molecular Formula C14H10O
Molecular Weight (g/mol) 194.23
MDL Number MFCD00001187
InChI Key RJGDLRCDCYRQOQ-UHFFFAOYSA-N
Synonym 9-10-dihydro-9-oxoanthracene, 9(10H)-Anthracenone
PubChem CID 7018
ChEBI CHEBI:33835
IUPAC Name 9,10-dihydroanthracen-9-one
SMILES O=C1C2=CC=CC=C2CC2=CC=CC=C12

Specifications

Formula Weight 194.22
Color Yellow
Quantity 100 g
Physical Form Powder
Chemical Name or Material Anthrone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.