missing translation for 'onlineSavingsMsg'
Learn More

Anthrone, ACS reagent

Product Code. 15159153 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic Bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15159153 500 g Plastic Bottle 500g
10488240 25 g Plastic Bottle 25g
10522164 100 g Plastic Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15159153 Supplier Thermo Scientific Acros Supplier No. 104965000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 90-44-8 | C14H10O | 194.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-44-8
Molecular Formula C14H10O
Molecular Weight (g/mol) 194.23
MDL Number MFCD00001187
InChI Key RJGDLRCDCYRQOQ-UHFFFAOYSA-N
Synonym anthrone, 9 10h-anthracenone, carbothrone, anthranone, 9-oxoanthracene, 9,10-dihydro-9-oxoanthracene, az-o, anthracen-9 10h-one, anthracene, 9,10-dihydro-9-oxo, unii-fp0fj7k744
PubChem CID 7018
ChEBI CHEBI:33835
IUPAC Name 10H-anthracen-9-one
SMILES O=C1C2=CC=CC=C2CC2=CC=CC=C12

Specifications

Absorbance 0.75 max. (425nm, 1cm cell) Passes Test, 0.045 max. (620nm, 1cm cell) Passes Test
Melting Point 152.0°C to 157.0°C
Color Yellow
Boiling Point 721.0°C
Infrared Spectrum Authentic
Packaging Plastic Bottle
Quantity 500 g
Beilstein 07,473
Sensitivity (to carbohydrates) Passes Test
Merck Index 15,682
Solubility Information (in ethyl acetate) Passes test
Formula Weight 194.23
Grade ACS Reagent
Physical Form Crystals
Chemical Name or Material Anthrone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.