Learn More
Anthranilic acid, 98+%
Product Code. p-7220484 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
5 kg
Unit Size:
100g
5kg
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
Product Code. 10182700
Brand: Thermo Scientific Acros 104911000
Request Bulk or Custom Format
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
CAS: 118-92-3 | C7;H7;NO2; | 137.14 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 118-92-3
Molecular Formula C7;H7;NO2;
Molecular Weight (g/mol) 137.14
Specifications
Melting Point 144.0°C to 148.0°C
Assay Percent Range 98+%
Packaging Plastic Bottle
Beilstein 14, 310
Merck Index 15, 417
Solubility Information Solubility aq. soln.: 5.7 g/L (25°C). Other solubilities: readily soluble in ethanol, esters, acetone, soluble in pyridine, hot chloroform, hot benzene
Density 1.4100 g/mL
Quantity 100 g
Flash Point 150°C
Formula Weight 137.14
Specific Gravity 1.4100
Chemical Name or Material Anthranilic acid
Show More Show Less
compliance-icons
Product Identifier
  • Anthranilic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.