accessibility menu, dialog, popup

UserName

CAS RN 118-92-3

NH2HOO

CAS RN 118-92-3

IUPAC Name: 2-aminobenzoic acid
Synonyms: 2-aminobenzoic acid monolithium salt anthranilic acid vitamin L ortho-amidobenzoic acid o-anthranilic acid monosodium salt anthranilic acid calcium (2:1) salt anthranilic acid ortho-amidobenzoate anthranilic acid, calcium (2:1) salt o-anthranilate
Molecular Weight (g/mol): 137.14
Molecular Formula: C7H7NO2
InChi Key: RWZYAGGXGHYGMB-UHFFFAOYSA-N
SMILES: NC1=CC=CC=C1C(O)=O
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results