missing translation for 'onlineSavingsMsg'
Learn More

Anthracene, Sigma-Aldrich™

Product Code. 15870751
Change view
Click to view available options
Quantity:
100 g
250 g
Unit Size:
100g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15870751 250 g 250g
15816630 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15870751 Supplier Sigma Aldrich Supplier No. 8201090250
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-12-7
Grade Research
Molecular Weight (g/mol) 178.23

Specifications

Melting Point 213°C to 217°C
Density 1.13 g/cm3 at 20°C
Boiling Point 340°C/1013 hPa
MDL Number MFCD00001240
Quantity 250 g
Synonym Anthracene, Paranaphthalene, Anthracene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.