missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Antennapedia Control Peptide
Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Description
The Antennapedia Control Peptide is composed of the protein transduction (PTD) sequence derived from antennapedia which renders the peptide cell permeable. It can be used as a control for peptides that contain the antennapedia PTD sequence. Source: Synthetic Amino Acid Sequence:DRQIKIWFQNRRMKWKK The Antennapedia Control Peptide has been validated for the following applications: Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
| Content And Storage | -20°Cor to +80°C. Avoid freeze-thaw cycles. |
| For Use With (Application) | Antigenic Blocking Peptide for NBP1-45000 |
| Product Type | Antennapedia Control Peptide |
| Quantity | 1 mg |
| Regulatory Status | RUO - research use only |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction