missing translation for 'onlineSavingsMsg'
Learn More

Aniline Hydrogen Phthalate, MP Biomedicals

Product Code. 11441785 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11441785 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11441785 Supplier MP Biomedicals Supplier No. 0520105610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 50930-79-5
Molecular Formula C14H13NO4
Quantity 10 g
Synonym aniline hydrogen phthalate, aniline phthalate, aniline; phthalic acid, 1,2-benzenedicarboxylic acid, compd. with benzenamine 1:1, c8h6o4.c6h7n, phthalic acid, compound with aniline 1:1, aniline hydrogenphthalate, aniline hydrogen phtalate, aniline phthalate solution, phthalic acid aniline salt
InChI Key QGPZXNSBZMHHSR-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)N.C1=CC=C(C(=C1)C(=O)O)C(=O)O
IUPAC Name aniline;phthalic acid
Molecular Weight (g/mol) 259.261
PubChem CID 170940
Formula Weight 259.26
Chemical Name or Material Aniline Hydrogen Phthalate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.