missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Ani 9
GREENER_CHOICE

Product Code. 18719238
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18719238 10 mg 10mg
18769228 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18719238 Supplier Tocris Bioscience™ Supplier No. 6076/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective TMEM16A (Anoctamin-1; ANO1) blocker

Ani 9 is a potent and selective TMEM16A (Anoctamin-1; ANO1) blocker (IC50 = 77 nM). Displays no effect on ANO2 activity (at 10 μM). Also exhibits no effect on intracellular calcium signaling or CFTR and ENaC channel activity (at 30 μM).
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ani 9
CAS 356102-14-2
Synonym 2-(4-Chloro-2-methylphenoxy)-N'-(2-methoxybenzylidene)acetohydrazide
Quantity 10 mg
Solubility Soluble to 100mM in DMSO
Molecular Formula C17H17ClN2O3
Formula Weight 332.78
Physical Form Solid
Color Off-white
Product Type Bioactive Small Molecule
Purity >98%
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.