missing translation for 'onlineSavingsMsg'
Learn More

Anhydrotetracycline Hydrochloride, TRC

Product Code. 30348183
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30348183 25 mg 25mg
30347841 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30348183 Supplier Toronto Research Chemicals Supplier No. TRCA65605025MG
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Anhydrotetracycline hydrochloride
CAS 13803-65-1
Purity Grade Notes HPLC
Molecular Formula C22H22N2O7.HCl
Quantity 25 mg
Synonym Anhydrotetracycline Hydrochloride, 2-Naphthacenecarboxamide, 4-(dimethylamino)-1,4,4a,5,12,12a-hexahydro-3,10,11,12a-tetrahydroxy-6-methyl-1,12-dioxo-, monohydrochloride, (4S,4aS,12aS)- (9CI), 2-Naphthacenecarboxamide, 4-(dimethylamino)-1,4,4a,5,12,12a-hexahydro-3,10,11,12a-tetrahydroxy-6-methyl-1,12-dioxo-, monohydrochloride (8CI), 2-Naphthacenecarboxamide, 4-(dimethylamino)-1,4,4a,5,12,12a-hexahydro-3,10,11,12a-tetrahydroxy-6-methyl-1,12-dioxo-, monohydrochloride, [4S-(4íÅ1⁄2±,4aíÅ1⁄2±,12aíÅ1⁄2±)]-, Chlortetracycline Hydrochloride Impurity J as Hydrochloride, Tetracycline Hydrochloride Impurity C as Hydrochloride, Chlortetracycline Impurity J as Hydrochloride, Tetracycline Impurity C as Hydrochloride
InChI Formula InChI=1S/C22H22N2O7.ClH/c1-8-9-5-4-6-12(25)13(9)17(26)14-10(8)7-11-16(24(2)3)18(27)15(21(23)30)20(29)22(11,31)19(14)28;/h4-6,11,16,25-27,31H,7H2,1-3H3,(H2,23,30);1H/t11-,16-,22-;/m0./s1
SMILES Cl.CN(C)[C@H]1[C@@H]2Cc3c(C)c4cccc(O)c4c(O)c3C(=O)[C@]2(O)C(=O)C(=C1O)C(=O)N
IUPAC Name (4S,4aS,12aS)-4-(dimethylamino)-3,10,11,12a-tetrahydroxy-6-methyl-1,12-dioxo-4a,5-dihydro-4H-tetracene-2-carboxamide;hydrochloride
Molecular Weight (g/mol) 462.88
Formula Weight 462.1194
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.