missing translation for 'onlineSavingsMsg'
Learn More

Andarine, TRC

Product Code. 18090310
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
2.50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18090310 5 mg 5mg
18072210 1 mg 1mg
18020410 2.5 mg 2.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18090310 Supplier Toronto Research Chemicals Supplier No. TRCA6374705MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 401900-40-1
Molecular Formula C19 H18 F3 N3 O6
Molecular Weight (g/mol) 441.36
Synonym (2S)-3-[4-(Acetylamino)phenoxy]-2-hydroxy-2-methyl-N-[4-nitro-3-(trifluoromethyl)phenyl]propanamide, Andarine, GTx 007, N-[4-Nitro-3-(trifluoromethyl)phenyl]-(2S)-3-[4-(acetylamino)phenoxy]-2-hydroxy-2-methylpropanamide, S 4, SARM S-4
IUPAC Name (2S)-3-(4-acetamidophenoxy)-2-hydroxy-2-methyl-N-[4-nitro-3-(trifluoromethyl)phenyl]propanamide
SMILES CC(=O)Nc1ccc(OC[C@](C)(O)C(=O)Nc2ccc(c(c2)C(F)(F)F)[N+](=O)[O-])cc1

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C19H18F3N3O6/c1-11(26)23-12-3-6-14(7-4-12)31-10-18(2,28)17(27)24-13-5-8-16(25(29)30)15(9-13)19(20,21)22/h3-9,28H,10H2,1-2H3,(H,23,26)(H,24,27)/t18-/m0/s1
Formula Weight 441.1148
Percent Purity >95%
Chemical Name or Material Andarine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.