Learn More
Andarine, TRC
Product Code. 18072210
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
2.50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18072210 1 mg 1mg
18090310 5 mg 5mg
18020410 2.5 mg 2.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 18072210 Supplier Toronto Research Chemicals Supplier No. TRCA6374701MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 401900-40-1
Molecular Formula C19 H18 F3 N3 O6
Molecular Weight (g/mol) 441.36
Synonym (2S)-3-[4-(Acetylamino)phenoxy]-2-hydroxy-2-methyl-N-[4-nitro-3-(trifluoromethyl)phenyl]propanamide, Andarine, GTx 007, N-[4-Nitro-3-(trifluoromethyl)phenyl]-(2S)-3-[4-(acetylamino)phenoxy]-2-hydroxy-2-methylpropanamide, S 4, SARM S-4
IUPAC Name (2S)-3-(4-acetamidophenoxy)-2-hydroxy-2-methyl-N-[4-nitro-3-(trifluoromethyl)phenyl]propanamide
SMILES CC(=O)Nc1ccc(OC[C@](C)(O)C(=O)Nc2ccc(c(c2)C(F)(F)F)[N+](=O)[O-])cc1
Specifications
Quantity 1 mg
InChI Formula InChI=1S/C19H18F3N3O6/c1-11(26)23-12-3-6-14(7-4-12)31-10-18(2,28)17(27)24-13-5-8-16(25(29)30)15(9-13)19(20,21)22/h3-9,28H,10H2,1-2H3,(H,23,26)(H,24,27)/t18-/m0/s1
Formula Weight 441.1148
Percent Purity >95%
Chemical Name or Material Andarine
Grade HPLC