Learn More
Abeba™ Anatom 32125 Shoes
Product Code. 15649974
Change view
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
48
49
50
51
52
Unit Size:
1 pair
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15649974 36 1 pair
15659974 37 1 pair
15669974 38 1 pair
15679974 39 1 pair
15689974 40 1 pair
15699974 41 1 pair
15609984 42 1 pair
15619984 43 1 pair
15629984 44 1 pair
15639984 45 1 pair
15649984 46 1 pair
15659984 47 1 pair
15669984 48 1 pair
15679984 49 1 pair
15689984 50 1 pair
15699984 51 1 pair
15609994 52 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
Product Code. 15649974 Supplier Abeba™ Supplier No. 32125/36
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
Resperation absorbing special lining with silver ions. Steel toe-cap.
  • CE EN ISO 20345:2011 SB A, E SRC
  • Microfiber black
  • Steel toe cap
  • Shock absorbant PU midsole
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3582)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Heel strap non-foldable, adjustable
  • Breathable lining with Silver ions
  • Instep area adjustable, with elastic band
  • velcro fastening
  • Suitable for heel and foot lift
  • Suitable for hygienically demanding areas
Specifications
Certifications/Compliance CE EN ISO 20345:2011 SB A, E SRC
Color Black
Material Microfiber
Size 36
Sole Features Slip-resistant
Sole Material Polyurethane
Style 32125
Toe Material Steel