missing translation for 'onlineSavingsMsg'
Learn More

Anastrozole, MedChemExpress

Product Code. 16496426
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16496426 100 mg 100mg
16482157 50 mg 50mg
16466018 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16496426 Supplier Medchem Express Supplier No. HY14274
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Anastrozole is a potent, highly selective aromatase inhibitor, which inhibits human placental aromatase with an IC50 of 15 nM.

Anastrozole is a potent, highly selective aromatase inhibitor, which inhibits human placental aromatase with an IC50 of 15 nM.

Specifications

Chemical Name or Material Anastrozole
Physical Form Solid
CAS 120511-73-1
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C17H19N5
For Use With (Application) Cancer-programmed cell death
Synonym ZD1033
Solubility Information DMSO : ≥ 100 mg/mL (340.87 mM)
SMILES CC(C)(C#N)C1=CC(C(C)(C#N)C)=CC(CN2C=NC=N2)=C1
Molecular Weight (g/mol) 293.37
Formula Weight 293.37
Percent Purity 99.54%
Grade Research
Afficher plus Afficher moins

Research purposes only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.