Learn More
OHAUS™ Analytical Balance - PR Series
Product Code. 16650324 Shop All Ohaus Products
Change view
Click to view available options
Model:
PR124M
PR224M
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Unit Size
16650324 PR224M Each
16640324 PR124M Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16650324 Supplier OHAUS™ Supplier No. 30524385
  • ONLINE EXCLUSIVE PRICE
    1725.00 EUR
    1288.00 EUR / Pack of 1
    Save 437.00 EUR
    25% Off
View all promotions
Estimated Shipment: 23-10-2026
Designed for Routine Weighing Applications in Your Workplace
  • The PR is equipped with 3 essential weighing modes, RS-232 connectivity for data transfer and storage, and internal calibration (in select models), making it ideal for routine weighing applications.
  • Equipped with an easy-to-read, bright backlit display and a simple user interface, the PR is incredibly easy to operate, with almost no training required.
  • The PR's small footprint saves desktop space while providing a large weighing surface. The PR is durably constructed, and features a stainless steel pan to withstand day-to-day use in the workplace.
TRUSTED_SUSTAINABILITY
Specifications
Calibration InCal™ - Semi Automatic
Calibration Points User-selectable span calibration points
Connection Type RS232 (one port included)
Description User-selectable environmental filters and brightness settings, auto-tare, auto-dim, user-selectable span calibration points, software lockout and reset menu, user-selectable communication settings and data print options, user-definable project and user IDs, software overload/underload indicator, stability indicator
Display Type Backlit LCD
Readability 0.0001 g
Humidity Range 80% RH
Linearity 0.0002 g
Model PR224M
Repeatability 0.0001 g
Stabilization Time 3.000 sec.
Weighing Modes Milligram, Gram, Carat
Dimensions (L x W x H) 317 x 201 x 303 mm
Length (English) 12.50 in.
Length (Metric) 317.00 mm
Width (English) 7.90 in.
Width (Metric) 201.00 mm
Height (English) 11.90 in.
Height (Metric) 303.000 mm
Depth (English) 12.50 in.
Depth (Metric) 317.00 mm
Weight (English) 10.000 lb.
Weight (Metric) 4.500 kg
Electrical Requirements AC Adapter (Included)