Learn More
Electrothermal™ Analogue Power Controller
Product Code. 10724216
Change view
Click to view available options
Voltage:
230 V
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Voltage Unit Size
10724216 230 V Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10724216 Supplier Electrothermal™ Supplier No. 3622302
  • ONLINE EXCLUSIVE PRICE
    334.00 EUR
    266.29 EUR / Pack of 1
    Save 67.71 EUR
    20% Off
Estimated Shipment: 07-12-2026
Control the power supply to one piece of laboratory equipment at a time. Analogue Power Controller is designed to control the heating of resistive loads for bench top operation.
  • Each heating controller has 2 neon indicators: ''Power On'' white neon light and ''Mantle/Bunsen Heater On'' amber neon light.
  • Regulator control knob can be turned clockwise to increase power.
  • As the knob is turned, the heating controller's amber neon light will pulsate to show that power is being supplied to the equipment being controlled, e.g. mantles, heating tape.
  • The pulse frequency decreases as the regulator control knob setting is increased
  • At maximum setting, the amber neon is continually illuminated.
  • Includes a support rod clamp at the rear of each heating controller to take a standard 12.5 mm (1/2in.) diameter rod.
  • Short mains lead with an IEC socket to connect to the resistive load.
  • An accessory extension mains lead is available where remote operation is required, e.g. in a fume extraction unit.
  • Mountable on standard 12 mm diameter scaffold or retort stands, and can also be wall mounted using a mounting bracket or stand directly on the bench.
Specifications
Controller Type Analogue
For Use With Heating Mantles
Dimensions (L x W x H) 130 x 95 x 105 mm
Voltage 230 V
Wattage 1800 W
Weight (Metric) 0.42 kg
Product Type Temperature Controller
Frequency 50/60 Hz