missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Amylase, Fungal, MP Biomedicals™

Product Code. 11413201 Shop All MP Biomedicals Products
Click to view available options
Quantity:
250 kU
Unit Size:
250ku
This item is not returnable. View return policy

Product Code. 11413201

Brand: MP Biomedicals™ 02150375.2

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Amylase, Fungal, MP Biomedicals is a fungal with >50ug/mg protein.

TRUSTED_SUSTAINABILITY

Specifications

Color Tan
pH Optimal pH range 4.8 to 5.5 (Lit.)
Content And Storage 0°C
Source Aspergillus oryzae
Quantity 250 kU
Form Powder
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.