missing translation for 'onlineSavingsMsg'
Learn More

Ampicillin, sodium salt, 99.67%, MP Biomedicals™

Product Code. 11440762 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11440762 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11440762 Supplier MP Biomedicals Supplier No. 0219419950

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Type Ampicillin Sodium Salt
Physical Form Powder
CAS 69-52-3
Color White
Quantity 50 mg
Assay Percent Range 0.9967
Molecular Formula C16H21N3NaO4S
Synonym D[-]-a-Aminobenzylpenicillin, 6-[D(-)-a-Aminophenylacetamido]penicillanic acid
InChI Key BSFVNXCYXDYHOD-ZQDFAFASSA-N
SMILES [HH].CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)N)C(=O)O)C.[Na]
IUPAC Name (2S,5R,6R)-6-[[(2R)-2-amino-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;molecular hydrogen;sodium
Molecular Weight (g/mol) 374.411
PubChem CID 131673879
Formula Weight 371.4
Chemical Name or Material Ampicillin, sodium salt
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.