missing translation for 'onlineSavingsMsg'
Learn More

AMOZ, TRC

Product Code. 30138466
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138466 25 mg 25mg
30136312 50 mg 50mg
17908704 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138466 Supplier Toronto Research Chemicals Supplier No. TRCA63460025MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 43056-63-9
Molecular Formula C8 H15 N3 O3
Molecular Weight (g/mol) 201.22
Synonym 2-Oxazolidinone, 3-amino-5-(morpholinomethyl)- (6CI,7CI), 3-Amino-5-(4-morpholinylmethyl)-2-oxazolidinone, 3-Amino-5-morpholinomethyl-2-oxazolidinone, 3-Amino-5-morpholinomethyl-2-oxazolidone, AMOZ
IUPAC Name 3-amino-5-(morpholin-4-ylmethyl)-1,3-oxazolidin-2-one
SMILES NN1CC(CN2CCOCC2)OC1=O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C8H15N3O3/c9-11-6-7(14-8(11)12)5-10-1-3-13-4-2-10/h7H,1-6,9H2
Formula Weight 201.1113
Percent Purity >95%
Chemical Name or Material 3-Amino-5-morpholinomethyl-1,3-oxazolidin-2-one
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.