missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Thiocyanate, MilliporeSigma™

Product Code. 15814060
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic Bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15814060 500 g Plastic Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15814060 Supplier Supelco Supplier No. 1012130500
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For analysis EMSURE™ ACS, ISO, Reag. Ph Eur

TRUSTED_SUSTAINABILITY

Specifications

CAS 1762-95-4
Physical Form Solid
Quantity 500 g
Molecular Formula CH4N2S
Synonym ammonium thiocyanate, ammoniumrhodanid, thiocyanic acid, ammonium salt, ammonium isothiocyanate, ammonium rhodanate, weedazol tl, trans-aid, ammonium sulfocyanate, ammonium sulfocyanide, rhodanine, ammonium salt
Solubility Information Soluble in water: 1600g/L at 20°C
InChI Key SOIFLUNRINLCBN-UHFFFAOYSA-N
SMILES C(#N)[S-].[NH4+]
IUPAC Name azanium;thiocyanate
Molecular Weight (g/mol) 76.117
PubChem CID 15666
Formula Weight 76.12
Percent Purity ≥99.0%
Grade ACS
Odor Odorless
Packaging Plastic Bottle
Vapor Pressure <1hPa at 20°C
Density 1.3g/cm3 at 20°C
Chemical Name or Material Ammonium Thiocyanate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.