Learn More
Ammonium tetrafluoroborate, 99.5% (metals basis)
Description
Ammonium tetrafluoroborate is used in the metal industry as a high-temperature flux. It is also used as a flame retardant and lubricant in aluminum cutting-oils. Further, it acts as an analytical reagent. In addition, it is used as a catalyst for resin finishing and also finds use in textile printing and dyeing industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 220°C (sublimation) |
| Color | White |
| Quantity | 250 g |
| UN Number | UN3260 |
| Solubility Information | Soluble in water. Insoluble in alcohol. |
| Formula Weight | 104.84 |
| Percent Purity | 99.5% |
| Odor | Ammonia-like |
| Sensitivity | Hygroscopic |
| Density | 1.871 g/mL |
| Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.