accessibility menu, dialog, popup

UserName

CAS RN 13826-83-0

BFFFFNH4

CAS RN 13826-83-0

IUPAC Name: ammonium tetrafluoroboranuide
Synonyms: ammonium tetrafluoroborate
Molecular Weight (g/mol): 104.84
Molecular Formula: BF4H4N
InChi Key: PDTKOBRZPAIMRD-UHFFFAOYSA-O
SMILES: [NH4+].F[B-](F)(F)F
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (2)
  • (3)
Physical Form 
  • (3)
  • (3)
  • (2)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (7)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (3)
  • (2)
  • (3)