missing translation for 'onlineSavingsMsg'
Learn More

Ammonium iron(III) hexacyanoferrate(II) hydrate, tech.

Product Code. p-7026103
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11444987 25 g 25g
11454987 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11444987 Supplier Thermo Scientific Alfa Aesar Supplier No. B21908.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 25869-00-5 | C6H4Fe2N7 | 285.837 g/mol

It is employed as a intermediate for pharmaceuticals. Also used in decorative cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediate for pharmaceuticals. Also used in decorative cosmetics.

Solubility
Soluble in water and mineral acids. Insoluble in alkali.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25869-00-5
Molecular Formula C6H4Fe2N7
Molecular Weight (g/mol) 285.837
MDL Number MFCD01726733
InChI Key ILZWGESBVHGTRX-UHFFFAOYSA-O
Synonym ammonium iron iii hexacyanoferrate ii, iron 3+ ammonium ferrocyanide, azanium;iron 2+ ;iron 3+ ;hexacyanide, ammonium iron iii hexacyanoferrate ii hydrate, ammonium iron iii hexacyanoferrate ii hydrate, tech., ammonium iron iii hexacyanoferrate ii giese salt
PubChem CID 23620040
IUPAC Name azanium;iron(2+);iron(3+);hexacyanide
SMILES [C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[NH4+].[Fe+2].[Fe+3]

Specifications

Quantity 25 g
Merck Index 14,7910
Formula Weight 285.83 (Anhydrous)
Grade Technical
Chemical Name or Material Ammonium iron(III) hexacyanoferrate(II) hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.