missing translation for 'onlineSavingsMsg'
Learn More

Ammonium iron(II) Sulfate Hexahydrate, Supelco™

Product Code. 15897190
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
Unit Size:
1kg
500g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15897190 1 kg 1kg
15844370 500 g 500g
15807200 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15897190 Supplier Supelco Supplier No. 1037921000
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For analysis EMSURE™ ISO

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-85-9
Grade Research
Molecular Weight (g/mol) 392.14

Specifications

Melting Point 39°C to 41°C (Decomposition)
Quantity 1 kg
MDL Number MFCD00150530
Synonym Ammonium iron(II) sulfate hexahydrate, Ammonium iron(II) sulfate hexahydrate
Density 1.86 g/cm3 at 20°C
Chemical Name or Material Ammonium Iron(II) Sulfate Hexahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.