missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Hydroxide, TRC

Product Code. 30008921
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008921 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30008921 Supplier Toronto Research Chemicals Supplier No. TRCA634020100ML
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1336-21-6
Molecular Formula H5NO
Synonym Ammonia Water, Ammonia, Aqua, Ammonia, Monohydrate, Aqua AmmoniaSX 1, SX 1 (Ammonia Water);
InChI Formula InChI=1S/H3N.H2O/h1H3;1H2
Quantity 100 mL
SMILES [NH4+].[OH-]
IUPAC Name azanium;hydroxide
Molecular Weight (g/mol) 35.05
Formula Weight 35.0371
Chemical Name or Material Ammonium Hydroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.