Learn More
Ammonium Dihydrogen Phosphate, MilliporeSigma™
Product Code. 15824030
Change view
Click to view available options
Quantity:
50 g
500 g
Packaging:
Fibre carton
Plastic Bottle
Unit Size:
500g
50kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15824030 500 g Plastic Bottle 500g
15834030 50 g Fibre carton 50kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15824030 Supplier Supelco Supplier No. 1011260500
Supelco Essentials Are in Stock
Use promo code 27958
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
For analysis EMSURE™ ACS, Reag. Ph Eur
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7722-76-1
Molecular Formula H6NO4P
Molecular Weight (g/mol) 115.03
MDL Number MFCD00003396
InChI Key LFVGISIMTYGQHF-UHFFFAOYSA-N
Synonym ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate
PubChem CID 24402
ChEBI CHEBI:62982
IUPAC Name phosphoric acid amine
SMILES N.OP(O)(O)=O
Specifications
Physical Form Solid
Quantity 500 g
Solubility Information Soluble in water: 368g/L at 20°C
Formula Weight 115.02
Percent Purity ≥99.0%
Grade ACS
Odor Odorless
Packaging Plastic Bottle
Vapor Pressure 0.066hPa at 125°C
Density 1.80g/cm3 at 20°C
Chemical Name or Material Ammonium dihydrogen phosphate
Show More Show Less
Product Identifier
  • Ammonium dihydrogen phosphate for analysis EMSURE® ACS,Reag. Ph Eur