missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Carbamate, Supelco™

Product Code. 15844030
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15844030 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
MQ300 Graded - For Products Used in Applications Requiring Enhanced Change Control and Quality Agreement
Product Code. 15844030 Supplier Supelco Supplier No. 1011340500
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
MQ300 Graded - For Products Used in Applications Requiring Enhanced Change Control and Quality Agreement

For analysis EMSURE™

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 152°C (Closed Capillary Tube)
CAS 1111-78-0
Quantity 500 g
MDL Number MFCD00013010
Synonym Ammonium carbamate, Ammonium carbaminate, Carbamic acid ammonium salt, Ammonium carbamate
Molecular Weight (g/mol) 78.07
Grade Research
Density 1.6 g/cm3 at 20°C
Chemical Name or Material Ammonium Carbamate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.