Learn More
Melford Ammonium Acetate
Description
Widely used reagent in molecular biology and chromatography.
Widely used reagent in molecular biology and chromatography.
Specifications
Specifications
| Chemical Name or Material | Ammonium Acetate |
| CAS | 631-61-8 |
| Quantity | 1 kg |
| Molecular Formula | C2H7NO2 |