missing translation for 'onlineSavingsMsg'
Learn More

Ammelide, TRC

Product Code. 30180321
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30180321 1 g 1g
30180066 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30180321 Supplier Toronto Research Chemicals Supplier No. TRCA6336501G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 645-93-2
Molecular Formula C3 H4 N4 O2
Molecular Weight (g/mol) 128.09
Synonym Ammelide (6CI), s-Triazine-2,4(1H,3H)-dione, 6-amino- (8CI), s-Triazine-2,4(1H,3H)-dione, dihydro-6-imino- (7CI), 6-Amino-1,3,5-triazine-2,4(1H,3H)-dione, 2-Amino-1,3,5-triazine-4,6-dione, 2-Amino-4,6-dihydroxy-s-triazine, Ammelid, GS 35713, Melanurenic acid, NSC 8146, s-Dihydroxyaminotriazine
IUPAC Name 6-amino-1,3,5-triazine-2,4-diol
SMILES Nc1nc(O)nc(O)n1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C3H4N4O2/c4-1-5-2(8)7-3(9)6-1/h(H4,4,5,6,7,8,9)
Formula Weight 128.0334
Percent Purity >95%
Chemical Name or Material Ammelide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.