Learn More
Amlodipine, TRC
Product Code. 18043920
Change view
Click to view available options
Quantity:
10 mg
100 mg
1 g
Unit Size:
100mg
10mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18043920 100 mg 100mg
17919829 10 mg 10mg
18074010 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 18043920 Supplier Toronto Research Chemicals Supplier No. TRCA633495100MG
 Restricted Item
Estimated Shipment: 02-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 88150-42-9
Molecular Formula C20 H25 Cl N2 O5
Molecular Weight (g/mol) 408.88
Synonym 2-[(2-aminoethoxy)methyl]-4-(2-chlorophenyl)-1,4-dihydro-6-methyl-3,5-Pyridinedicarboxylic acid 3-ethyl 5-methyl ester, (R,S)-Amlodipine, 2-[(2-Aminoethoxy)methyl]-4-(2-chlorophenyl)-3-(ethoxycarbonyl)-5-(methoxycarbonyl)-6-methyl-1,4-dihydropyridine, Amlodipine, Pelmec, Racemic Amlodipine
IUPAC Name 3-O-ethyl 5-O-methyl 2-(2-aminoethoxymethyl)-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate
SMILES CCOC(=O)C1=C(COCCN)NC(=C(C1c2ccccc2Cl)C(=O)OC)C
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C20H25ClN2O5/c1-4-28-20(25)18-15(11-27-10-9-22)23-12(2)16(19(24)26-3)17(18)13-7-5-6-8-14(13)21/h5-8,17,23H,4,9-11,22H2,1-3H3
Formula Weight 408.1452
Percent Purity >95%
Chemical Name or Material Amlodipine
Grade HPLC