Learn More
Amlodipine Mesilate, Mikromol™
Description
Amlodipine Mesilate
Specifications
Specifications
| Chemical Name or Material | Amlodipine Mesilate |
| CAS | 246852-12-0 |
| Analyte or Component Names | Amlodipine Mesilate |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C20 H25 Cl N2 O5 . C H4 O3 S |
| Certifications/Compliance | ISO 17025 |
| Synonym | Amlodipine Mesilate |
| InChI Formula | InChI=1S/C20H25ClN2O5.CH4O3S/c1-4-28-20(25)18-15(11-27-10-9-22)23-12(2)16(19(24)26-3)17(18)13-7-5-6-8-14(13)21;1-5(2,3)4/h5-8,17,23H,4,9-11,22H2,1-3H3;1H3,(H,2,3,4) |
| SMILES | CCOC(=O)C1=C(COCCN)NC(=C(C1c2ccccc2Cl)C(=O)OC)C.CS(=O)(=O)O |
| IUPAC Name | 3-O-ethyl 5-O-methyl 2-(2-aminoethoxymethyl)-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate;methanesulfonic acid |
| Show More |