Learn More
Aminophylline, MedChemExpress
Product Code. 16488126
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16488126 100 mg 100mg
16402046 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16488126 Supplier Medchem Express Supplier No. HYB0140
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
MedChemExpress Aminophylline is a competitive and non-selective phosphodiesterase (PDE) inhibitor. Aminophylline is a competitive adenosine receptor antagonist. Aminophylline has apulmonary vasodilator action as well as a bronchodilator action and has the potential for asthma research.
Aminophylline is a competitive and non-selective phosphodiesterase (PDE) inhibitor. Aminophylline is a competitive adenosine receptor antagonist. Aminophylline has apulmonary vasodilator action as well as a bronchodilator action and has the potential for asthma research.
Specifications
Chemical Name or Material Aminophylline
Physical Form Solid
CAS 317-34-0
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C16H24N10O4
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 14.29 mg/mL (33.99 mM; Need ultrasonic) ∣H2O : 6.25 mg/mL (14.87 mM; Need ultrasonic)
SMILES O=C(N1C)N(C)C2=C(N=CN2)C1=O.O=C(N3C)N(C)C4=C(N=CN4)C3=O.NCCN
Molecular Weight (g/mol) 420.43
Formula Weight 420.43
Percent Purity 99.91%
Grade Research