missing translation for 'onlineSavingsMsg'
Learn More

Aminophenyltrimethoxysilane, TRC

Product Code. 30179494
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179494 500 mg 500mg
30179011 50 mg 50mg
30179140 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179494 Supplier Toronto Research Chemicals Supplier No. TRCA638108500MG
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33976-43-1
Molecular Formula C9H15NO3Si
Molecular Weight (g/mol) 213.31
Synonym 4-(Trimethoxysilyl)benzenamine, (4-Aminophenyl)trimethoxysilane, (p-Aminophenyl)trimethoxysilane, 4-(Trimethoxysilyl)aniline, A 0724, p-(Trimethoxysilyl)aniline, gamma-Anilinotrimethoxysilane
IUPAC Name 4-trimethoxysilylaniline
SMILES CO[Si](OC)(OC)c1ccc(N)cc1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C9H15NO3Si/c1-11-14(12-2,13-3)9-6-4-8(10)5-7-9/h4-7H,10H2,1-3H3
Formula Weight 213.0821
Chemical Name or Material Aminophenyltrimethoxysilane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.