missing translation for 'onlineSavingsMsg'
Learn More

δ-Aminolevulinic acid hydrochloride, 99.7%, MP Biomedicals™

Product Code. 11422910 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
0.1 g
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11422910 500 mg 500mg
11412910 0.1 g 100mg
11432910 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11422910 Supplier MP Biomedicals Supplier No. 0210024790
  • ONLINE EXCLUSIVE PRICE
    149.00 EUR
    141.75 EUR / 500mg
    Save 7.25 EUR
    4% Off
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

delta-Aminolevulinic acid hydrochloride is the universal precursor of tetrapyrroles, such as chlorophyll and heme

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9-2-5451
Molecular Formula C5H10ClNO3
Molecular Weight (g/mol) 167.59
MDL Number MFCD00012869
InChI Key ZLHFONARZHCSET-UHFFFAOYSA-N
Synonym 5-aminolevulinic acid hydrochloride, 5-amino-4-oxopentanoic acid hydrochloride, aminolevulinic acid hydrochloride, levulan, levulan kerastick, aminolevulinic acid hcl, delta-aminolevulinic acid hydrochloride, gliolan, ameluz, 5-aminolevulinate hydrochloride
PubChem CID 123608
IUPAC Name 5-amino-4-oxopentanoic acid;hydrochloride
SMILES C(CC(=O)O)C(=O)CN.Cl

Specifications

Color White
Assay Percent Range 0.997
Quantity 500 mg
Formula Weight 167.6
Physical Form Crystalline Powder
Chemical Name or Material δ-Aminolevulinic acid hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.