Learn More
Aminohippurate sodium, MedChemExpress
Product Code. 16408026
Encompass_Preferred
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. 16408026
Brand: Medchem Express HYA0080
MedChemExpress Aminohippurate sodium is a diagnostic agent useful in medical tests involving the kidney used in the measurement of renal plasma flow.
Aminohippurate sodium is a diagnostic agent useful in medical tests involving the kidney used in the measurement of renal plasma flow.
Specifications
Chemical Name or Material Aminohippurate sodium
Physical Form Solid
CAS 94-16-6
Color White to Light Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C9H9N2NaO3
Synonym Sodium p-aminohippurate p-Aminohippuric acid sodium salt
Solubility Information H2O : 100 mg/mL (462.60 mM; Need ultrasonic) ∣DMSO : ≥ 46 mg/mL (212.80 mM)
SMILES O=C(O[Na])CNC(C1=CC=C(N)C=C1)=O
Molecular Weight (g/mol) 216.17
Formula Weight 216.17
Percent Purity 99.96%
Grade Research