Learn More
Aminoglutethimide, LoGiCal
Product Code. 30462724
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. Quantity unitSize
30462724 10 mg 10mg
1 options
Product Code. 30462724
Brand: LoGiCal LGCFOR1777.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Aminoglutethimide
CAS 125-84-8
Analyte or Component Names Aminoglutethimide
Shipping Condition Room Temperature
Molecular Formula C13 H16 N2 O2
Certifications/Compliance ISO 9001
Synonym Glutarimide, 2-(p-aminophenyl)-2-ethyl- (6CI,7CI,8CI), (RS)-Aminoglutethimide, 2-(p-Aminophenyl)-2-ethylglutarimide, 3-(4-Aminophenyl)-3-ethylpiperidine-2,6-dione, Aminoglutethimide, Ba-16038, Cytadren, DL-Aminoglutethimide, Elipten, NSC 330915, Orimeten, dl-Aminoglutethimide, p-Aminoglutethimide, 2,6-Piperidinedione, 3-(4-aminophenyl)-3-ethyl-, (+/-)-Aminoglutethimide, alpha-(p-Aminophenyl)-alpha-ethylglutarimide
InChI Formula InChI=1S/C13H16N2O2/c1-2-13(8-7-11(16)15-12(13)17)9-3-5-10(14)6-4-9/h3-6H,2,7-8,14H2,1H3,(H,15,16,17)
Solution Type Neat
SMILES CCC1(CCC(=O)NC1=O)c2ccc(N)cc2
IUPAC Name 3-(4-aminophenyl)-3-ethylpiperidine-2,6-dione
Industry Type Clinical Testing
Molecular Weight (g/mol) 232.28
Quantity 10 mg
Formula Weight 232.1212 g/mol