Learn More
Aminoacyl tRNA synthetase-IN-1, MedChemExpress
Product Code. 16461119
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16461119 1 mg 1mg
16456967 5 mg 5mg
16434248 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16461119 Supplier Medchem Express Supplier No. HY108939
 Restricted Item
Estimated Shipment: 02-11-2026
MedChemExpress Aminoacyl tRNA synthetase-IN-1 is a bacterial aminoacyl tRNA synthetase (aaRS) inhibitor.
Aminoacyl tRNA synthetase-IN-1 is a bacterial aminoacyl tRNA synthetase (aaRS) inhibitor.
Specifications
Chemical Name or Material Aminoacyl tRNA synthetase-IN-1
CAS 219931-45-0
Color White
Purity Grade Notes Research
Molecular Formula C16H25N7O7S
Quantity 1 mg
Solubility Information DMSO : 65 mg/mL (141.46 mM; Need ultrasonic)
SMILES O[C@@H]1[C@@H](COS(=O)(NC([C@H]([C@@H](C)CC)N)=O)=O)O[C@@H](N2C(N=CN=C3N)=C3N=C2)[C@@H]1O
Molecular Weight (g/mol) 459.48
Formula Weight 459.48
Percent Purity 99.63%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation