missing translation for 'onlineSavingsMsg'
Learn More

Aminoacetonitrile Sulfate, Sigma-Aldrich™

Product Code. 15814721
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15814721 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15814721 Supplier Sigma Aldrich Supplier No. 8410340050
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Specifications

CAS 5466-22-8
Melting Point 118°C to 122°C
MDL Number MFCD01941514
Quantity 50 g
Synonym Aminoacetonitrile sulfate, Glycinonitrile sulphate, Aminoacetonitrile sulfate
Molecular Weight (g/mol) 210.21
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.