Learn More
MP Biomedicals Amino Acid Diet, Synthetic, Powder
Product Code. 11437341 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 kg
20 kg
50 kg
Unit Size:
10kg
20kg
50kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11437341 10 kg 10kg
11447341 20 kg 20kg
11457341 50 kg 50kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11437341 Supplier MP Biomedicals Supplier No. 0296036110
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Diet rich in Amino Acids. Can be added into agar gel to increase palatability.
  • Diet rich in amino acids
  • Can be added into agar gel to increase palatability
  • Can be used for research
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Product Type Animal Diet
Disposable Single-use
Format Powder
Quantity 10 kg
Sterility Non-sterile
Storage Requirements Refrigerated
Length (English) 1 ft.
Length (Metric) 0.3048 m
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.