Learn More
Amifampridine, MedChemExpress
Description
Amifampridine (3,4-Diaminopyridine) is an orally active potassium channel blocker (PCB), can block presynaptic potassium channels. Amifampridine can be used in Lambert-Eaton myasthenic syndrome (LEMS) research.
Specifications
Specifications
| Chemical Name or Material | Amifampridine |
| CAS | 54-96-6 |
| Color | Gray |
| Purity Grade Notes | Research |
| Molecular Formula | C5H7N3 |
| Quantity | 100 mg |
| Synonym | 3,4-Diaminopyridine |
| Solubility Information | DMSO : ≥ 38 mg/mL (348.21 mM) |
| SMILES | NC1=C(N)C=CN=C1 |
| Molecular Weight (g/mol) | 109.13 |
| Show More |