Learn More
Thermo Scientific™ Amies Agar Gel Mini Tip: Firm Applicator Wire
Product Code. 10260483
Change view
Click to view available options
Quantity:
50/Pk., 500/Cs.
Unit Size:
Pack of 500
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10260483 50/Pk., 500/Cs. Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10260483 Supplier Thermo Scientific™ Supplier No. TS0012A
  • ONLINE EXCLUSIVE PRICE
    742.00 EUR
    656.66 EUR / Pack of 500
    Save 85.34 EUR
    11% Off
 Restricted Item
Estimated Shipment: 04-01-2027
Simplify the transportation of eye, nasopharyngeal, male urethral swab and pediatric swabs with these Amies Agar Gel Transport Swabs.
Simplify the transportation of eye, nasopharyngeal, male urethral swab and pediatric sampling with Thermo Scientific™ Oxoid™ Amies Agar Gel Mini Tip: Firm Applicator Wire. The narrow dimension of the swab shaft and tip provides a more practical device for some sampling.
Ensure optimal sample collection and recovery using Amies Agar Gel Mini Tip: Firm Applicator Wire.
  • Contains media for the maintenance of bacterial sample during transport to the laboratory.
  • Each batch of product is tested for performance using a wide range of aerobic and anaerobic pathogens to ensure adequate recoveries.
  • Lot number/expiry date and tamper-evident tube ensure complete traceability and sterility.
  • 'Venturi' Hour-Glass Tube Design prevents disintegration of gel column and eliminates undesirable air pockets, bubbles and breaks in gel which may be harmful to fastidious bacteria
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Transport Swab
Description Amies Agar Gel Mini Tip: Firm Applicator Wire
Quantity 50/Pk., 500/Cs.
Shaft Material Aluminum Wire
Storage Requirements 5°C to 25C°; do not freeze
Tip Material Rayon
Tip Type Minitip